accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 716-39-2

IUPAC Name: 1H,3H-naphtho[2,3-c]furan-1,3-dione
Synonyms: naphtho[2,3-c]furan-1,3-dione
Molecular Weight (g/mol): 198.18
Molecular Formula: C12H6O3
InChi Key: IZJDCINIYIMFGX-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=C1C=C1C=CC=CC1=C2
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)