accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

Cl

CAS RN 716-53-0

IUPAC Name: 9-chloroanthracene
Synonyms: 9-chloroanthracene
Molecular Weight (g/mol): 212.68
Molecular Formula: C14H9Cl
InChi Key: KULLJOPUZUWTMF-UHFFFAOYSA-N
SMILES: ClC1=C2C=CC=CC2=CC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)