accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClSH2N

CAS RN 71625-90-6

IUPAC Name: 1-(5-chloro-1-benzothiophen-3-yl)methanamine
Synonyms: 1-(5-chloro-1-benzothiophen-3-yl)methanamine
Molecular Weight (g/mol): 197.68
Molecular Formula: C9H8ClNS
InChi Key: VRNXLYAXYIHHHH-UHFFFAOYSA-N
SMILES: NCC1=CSC2=CC=C(Cl)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results