CAS RN 71802-31-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Arsenocholine Bromide, TRC
CAS: 71802-31-8 Molecular Formula: C5 H14 As O . Br Molecular Weight (g/mol): 244.99 Synonym: (2-Hydroxyethyl)trimethylarsonium Bromide; IUPAC Name: 2-hydroxyethyl(trimethyl)arsanium;bromide SMILES: [Br-].C[As+](C)(C)CCO