accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OSONNNN

CAS RN 7189-69-7

IUPAC Name: 1-(1H-imidazole-1-sulfonyl)-1H-imidazole
Synonyms: 1-(imidazole-1-sulfonyl)imidazole
Molecular Weight (g/mol): 198.2
Molecular Formula: C6H6N4O2S
InChi Key: ZLKNPIVTWNMMMH-UHFFFAOYSA-N
SMILES: O=S(=O)(N1C=CN=C1)N1C=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results