accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 71989-26-9

HN(S)OOOOOHNOHH3CH3CCH3HCl

CAS RN 71989-26-9

IUPAC Name: 6-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: 6-[(tert-butoxycarbonyl)amino]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol): 468.55
Molecular Formula: C26H32N2O6
InChi Key: UMRUUWFGLGNQLI-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NCCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)