accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 71989-93-0

IUPAC Name: 2,4,6-trihydroxybenzoate
Synonyms: 2,4,6-trihydroxybenzoate
Molecular Weight (g/mol): 169.11
Molecular Formula: C7H5O5
InChi Key: IBHWREHFNDMRPR-UHFFFAOYSA-M
SMILES: OC1=CC(O)=C(C([O-])=O)C(O)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results