accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 72-17-3

IUPAC Name: sodium 2-hydroxypropanoate
Synonyms: sodium lactate
Molecular Weight (g/mol): 112.06
Molecular Formula: C3H5NaO3
InChi Key: NGSFWBMYFKHRBD-UHFFFAOYNA-M
SMILES: [Na+].CC(O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results