accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C(S)OHONH2CH3

CAS RN 72-18-4

IUPAC Name: 2-amino-3-methylbutanoic acid
Synonyms: valin valina Hval
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: KZSNJWFQEVHDMF-UHFFFAOYNA-N
SMILES: CC(C)C(N)C(O)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (1)
  • (4)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (1)
  • (7)
Grade 
  • (5)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (8)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (1)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (7)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)