accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOFFF

CAS RN 720-94-5

IUPAC Name: 4,4,4-trifluoro-1-(4-methylphenyl)butane-1,3-dione
Synonyms: 4,4,4-trifluoro-1-(4-methylphenyl)butane-1,3-dione
Molecular Weight (g/mol): 230.19
Molecular Formula: C11H9F3O2
InChi Key: WRZMHTIRFOFFPY-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)CC(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results