accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOONOOCH3

CAS RN 72030-81-0

IUPAC Name: 4,5-dimethyl 1,3-oxazole-4,5-dicarboxylate
Synonyms: 4,5-dimethyl 1,3-oxazole-4,5-dicarboxylate
Molecular Weight (g/mol): 185.14
Molecular Formula: C7H7NO5
InChi Key: LYRDBSFJJDATFO-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N=CO1)C(=O)OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results