accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7205-98-3

IUPAC Name: chloromethanesulfonylbenzene
Synonyms: chloromethyl phenyl sulfone
Molecular Weight (g/mol): 190.64
Molecular Formula: C7H7ClO2S
InChi Key: NXAIQSVCXQZNRY-UHFFFAOYSA-N
SMILES: ClCS(=O)(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results