accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NCH3HOH3C(Z)

CAS RN 7207-49-0

IUPAC Name: (E)-N-(octan-2-ylidene)hydroxylamine
Synonyms: (E)-N-(octan-2-ylidene)hydroxylamine
Molecular Weight (g/mol): 143.23
Molecular Formula: C8H17NO
InChi Key: GZRPVYSKBVDCBV-CMDGGOBGSA-N
SMILES: CCCCCC\C(C)=N\O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results