accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNH2CH3

CAS RN 72072-16-3

IUPAC Name: 4-butyl-2-methylaniline
Synonyms: 4-butyl-2-methylaniline
Molecular Weight (g/mol): 163.26
Molecular Formula: C11H17N
InChi Key: JTXOXRXZCAMPHL-UHFFFAOYSA-N
SMILES: CCCCC1=CC(C)=C(N)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results