accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7209-38-3

IUPAC Name: 3-[4-(3-aminopropyl)piperazin-1-yl]propan-1-amine
Synonyms: 1,4-piperazinedipropanamine
Molecular Weight (g/mol): 200.33
Molecular Formula: C10H24N4
InChi Key: XUSNPFGLKGCWGN-UHFFFAOYSA-N
SMILES: NCCCN1CCN(CCCN)CC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results