accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOFFF

CAS RN 721-63-1

IUPAC Name: ethyl 2-[4-(trifluoromethyl)phenyl]acetate
Synonyms: ethyl 2-[4-(trifluoromethyl)phenyl]acetate
Molecular Weight (g/mol): 232.2
Molecular Formula: C11H11F3O2
InChi Key: BDVKGYOFECBKDX-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=C(C=C1)C(F)(F)F
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results