CAS RN 72178-02-0
IUPAC Name:
sodium {5-[2-chloro-4-(trifluoromethyl)phenoxy]-2-nitrobenzoyl}(methanesulfonyl)azanide
Synonyms:
sodium 5-[2-chloro-4-(trifluoromethyl)phenoxy]-2-nitrobenzoyl(methanesulfonyl)azanide
Molecular Weight (g/mol):
460.74
Molecular Formula:
C15H9ClF3N2NaO6S
InChi Key:
CRHGSCXKJPJNAB-UHFFFAOYSA-M
SMILES:
[Na+].CS(=O)(=O)[N-]C(=O)C1=CC(OC2=CC=C(C=C2Cl)C(F)(F)F)=CC=C1[N+]([O-])=O
1
–
1
of
1
results
1
Fomesafen, TRC
CAS: 72178-02-0 Molecular Formula: C15 H10 Cl F3 N2 O6 S Molecular Weight (g/mol): 438.76 Synonym: 5-[2-Chloro-4-(trifluoromethyl)phenoxy]-N-(methylsulfonyl)-2-nitrobenzamide,Flex,Flexstar,Fomesafen,N-Methanesulfonyl-5-[(2-chloro-α,α,α-trifluoro-4-tolyl)oxy]-2-nitrobenzamide,PP021,Reflex,Reflex (herbicide) IUPAC Name: 5-[2-chloro-4-(trifluoromethyl)phenoxy]-N-methylsulfonyl-2-nitrobenzamide SMILES: CS(=O)(=O)NC(=O)c1cc(Oc2ccc(cc2Cl)C(F)(F)F)ccc1[N+](=O)[O-]