accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 72198-83-5

IUPAC Name: 3-methyl-4-nitrobenzohydrazide
Synonyms: 3-methyl-4-nitrobenzohydrazide
Molecular Weight (g/mol): 195.18
Molecular Formula: C8H9N3O3
InChi Key: SMRIMKXHORAHJR-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1[N+]([O-])=O)C(=O)NN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results