accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrBrSOOCl

CAS RN 72256-95-2

IUPAC Name: 2,4-dibromobenzene-1-sulfonyl chloride
Synonyms: 2,4-dibromobenzenesulfonyl chloride
Molecular Weight (g/mol): 334.41
Molecular Formula: C6H3Br2ClO2S
InChi Key: OMOXIKAWVFDFNX-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=C(Br)C=C(Br)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)