accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3HNOHNONH2CH3OCH3OONHCH3OOH3CCH3NH3CONH3CONONHOCH3CH3NNH2CH3OOCH3CH3NH3CONH3CONO

CAS RN 7240-37-1

IUPAC Name: 2,7-diamino-4,6-dimethyl-3-oxo-N1,N9-bis[2,5,9-trimethyl-1,4,7,11,14-pentaoxo-6,13-bis(propan-2-yl)-hexadecahydro-1H-pyrrolo[2,1-i]1-oxa-4,7,10,13-tetraazacyclohexadecan-10-yl]-3H-phenoxazine-1,9-dicarboxamide
Synonyms: 7-aminoactinomycin D
Molecular Weight (g/mol): 1270.45
Molecular Formula: C62H87N13O16
InChi Key: YXHLJMWYDTXDHS-UHFFFAOYNA-N
SMILES: CC(C)C1NC(=O)C(NC(=O)C2=C3N=C4C(OC3=C(C)C(N)=C2)=C(C)C(=O)C(N)=C4C(=O)NC2C(C)OC(=O)C(C(C)C)N(C)C(=O)CN(C)C(=O)C3CCCN3C(=O)C(NC2=O)C(C)C)C(C)OC(=O)C(C(C)C)N(C)C(=O)CN(C)C(=O)C2CCCN2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results