accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7240-90-6

IUPAC Name: X-gal
Synonyms: X-gal
Molecular Weight (g/mol): 594.7
Molecular Formula: C23H41Cu2N5O5
InChi Key: GGILAJWFDJWIQO-UHFFFAOYSA-N
SMILES: [Cu+].[Cu++].CC(O)O.CC(O)O.CN1CCN(CC2=CC(=CC(CN3CCN(C)CC3)=C2O)C#N)CC1
Molecular Weight (g/mol) 
  • (15)
  • (9)
Quantity 
  • (4)
  • (1)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
Physical Form 
  • (6)
  • (10)
  • (2)
Percent Purity 
  • (3)
  • (5)
  • (2)
  • (5)
  • (2)
Grade 
  • (2)
  • (3)
  • (1)
  • (4)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (6)
  • (5)
  • (3)
  • (3)
  • (15)
  • (9)
  • (4)
  • (1)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (6)
  • (10)
  • (2)
  • (3)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (4)
  • (5)