CAS RN 72482-64-5
IUPAC Name:
2,4-difluorobenzoyl chloride
Synonyms:
2,4-difluorobenzoyl chloride
Molecular Weight (g/mol):
176.55
Molecular Formula:
C7H3ClF2O
InChi Key:
JSWRVDNTKPAJLB-UHFFFAOYSA-N
SMILES:
FC1=CC=C(C(Cl)=O)C(F)=C1
1
–
1
of
1
results
1
2,4-Difluorobenzoyl chloride, 97%
CAS: 72482-64-5 Molecular Formula: C7H3ClF2O Molecular Weight (g/mol): 176.55 MDL Number: MFCD00000658 InChI Key: JSWRVDNTKPAJLB-UHFFFAOYSA-N PubChem CID: 588081 IUPAC Name: 2,4-difluorobenzoyl chloride SMILES: FC1=CC=C(C(Cl)=O)C(F)=C1