accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNN

CAS RN 7251-61-8

IUPAC Name: 2-methylquinoxaline
Synonyms: 2-methylquinoxaline
Molecular Weight (g/mol): 144.18
Molecular Formula: C9H8N2
InChi Key: ALHUXMDEZNLFTA-UHFFFAOYSA-N
SMILES: CC1=CN=C2C=CC=CC2=N1
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (3)
Boiling Point 
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)