accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClH2N

CAS RN 7252-53-1

IUPAC Name: 1-cyclopropylmethanamine hydrochloridyl
Synonyms: 1-cyclopropylmethanamine hydrochloridyl
Molecular Weight (g/mol): 106.57
Molecular Formula: C4H9ClN
InChi Key: MQCZBGGAZQXTLH-UHFFFAOYSA-N
SMILES: [Cl].NCC1CC1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results