accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 72556-74-2

H3CNCH3H3COPOOHHOH2OH2OH2OH2OClCa

CAS RN 72556-74-2

IUPAC Name: [2-(trimethylazaniumyl)ethoxy]phosphonic acid calcium tetrahydrate chloride
Synonyms: phosphocholine calcium tetrahydrate chloride
Molecular Weight (g/mol): 331.74
Molecular Formula: C5H23CaClNO8P
InChi Key: YFRZEXLBVHTMGV-UHFFFAOYSA-N
SMILES: O.O.O.O.[Cl-].[Ca].C[N+](C)(C)CCOP(O)(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results