accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 72745-76-7

IUPAC Name: 2-methyl-1,3-benzoxazol-5-amine
Synonyms: 2-methyl-1,3-benzoxazol-5-amine
Molecular Weight (g/mol): 148.17
Molecular Formula: C8H8N2O
InChi Key: FECYTFWPNCBIHC-UHFFFAOYSA-N
SMILES: CC1=NC2=CC(N)=CC=C2O1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results