accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOFF

CAS RN 72768-94-6

IUPAC Name: [2-(difluoromethoxy)phenyl]methanol
Synonyms: [2-(difluoromethoxy)phenyl]methanol
Molecular Weight (g/mol): 174.15
Molecular Formula: C8H8F2O2
InChi Key: JJAUYEZYAXAERE-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1OC(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results