accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 72784-42-0

IUPAC Name: methyl 1-aminocyclopropane-1-carboxylate hydrochloride
Synonyms: methyl 1-aminocyclopropane-1-carboxylate hydrochloride
Molecular Weight (g/mol): 151.59
Molecular Formula: C5H10ClNO2
InChi Key: CHSDCWKLSHPKFY-UHFFFAOYSA-N
SMILES: Cl.COC(=O)C1(N)CC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results