accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 72846-00-5

IUPAC Name: 1-benzyl-5-phenyl-1,3-diazinane-2,4,6-trione
Synonyms: 1-benzyl-5-phenyl-1,3-diazinane-2,4,6-trione
Molecular Weight (g/mol): 294.31
Molecular Formula: C17H14N2O3
InChi Key: KCWWCWMGJOWTMY-UHFFFAOYNA-N
SMILES: O=C1NC(=O)N(CC2=CC=CC=C2)C(=O)C1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results