accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3NNCH3H3CCH3

CAS RN 72914-19-3

IUPAC Name: 4,4'-di-tert-butyl-2,2'-bipyridine
Synonyms: 4,4'-di-tert-butyl-2,2'-bipyridine
Molecular Weight (g/mol): 268.4
Molecular Formula: C18H24N2
InChi Key: TXNLQUKVUJITMX-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=NC=C1)C1=NC=CC(=C1)C(C)(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results