accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH(S)ONH2OH

CAS RN 73-22-3

IUPAC Name: 2-amino-3-(1H-indol-3-yl)propanoic acid
Synonyms: optimax ardeytropin ardeydorm trofan triptofano indole-3-alanine pharmascience brand of tryptophan DL-trytophane tryptophan metabolism alterations DL tryptophan
Molecular Weight (g/mol): 204.23
Molecular Formula: C11H12N2O2
InChi Key: QIVBCDIJIAJPQS-UHFFFAOYNA-N
SMILES: NC(CC1=CNC2=CC=CC=C12)C(O)=O
Quantity 
  • (5)
  • (2)
  • (1)
  • (5)
  • (5)
  • (1)
  • (3)
Percent Purity 
  • (4)
  • (3)
  • (2)
  • (8)
Grade 
  • (1)
  • (3)
  • (1)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (5)
  • (6)
  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (5)
  • (5)
  • (1)
  • (3)
  • (4)
  • (3)
  • (2)
  • (8)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)