accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NOHNNHN

CAS RN 73-40-5

IUPAC Name: 2-amino-6,7-dihydro-3H-purin-6-one
Synonyms: 2-aminohypoxanthine
Molecular Weight (g/mol): 151.13
Molecular Formula: C5H5N5O
InChi Key: UYTPUPDQBNUYGX-UHFFFAOYSA-N
SMILES: NC1=NC(=O)C2=C(N1)N=CN2
Quantity 
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (4)
  • (1)