accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NOHH2NN

CAS RN 73-67-6

IUPAC Name: (4-amino-2-methylpyrimidin-5-yl)methanol
Synonyms: toxopyrimidine
Molecular Weight (g/mol): 139.16
Molecular Formula: C6H9N3O
InChi Key: VUTBELPREDJDDH-UHFFFAOYSA-N
SMILES: CC1=NC=C(CO)C(N)=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results