accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOFNOO

CAS RN 7304-32-7

IUPAC Name: 2-fluoro-5-nitrobenzoic acid
Synonyms: 2-fluoro-5-nitrobenzoic acid
Molecular Weight (g/mol): 185.11
Molecular Formula: C7H4FNO4
InChi Key: ICXSHFWYCHJILC-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC=C1F)[N+]([O-])=O
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results