accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 73075-45-3

IUPAC Name: 7-chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride
Synonyms: 7-chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride
Molecular Weight (g/mol): 204.09
Molecular Formula: C9H11Cl2N
InChi Key: OGIAIXMUSSACDB-UHFFFAOYSA-N
SMILES: Cl.ClC1=CC2=C(CCNC2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results