accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOSBr

CAS RN 7311-64-0

IUPAC Name: 3-bromothiophene-2-carboxylate
Synonyms: 3-bromothiophene-2-carboxylate
Molecular Weight (g/mol): 206.03
Molecular Formula: C5H2BrO2S
InChi Key: VQQLWBUTTMNMFT-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=C(Br)C=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results