accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

Mn

CAS RN 73138-26-8

IUPAC Name: manganocene
Synonyms: manganocene
Molecular Weight (g/mol): 180.09
Molecular Formula: C10H5Mn
InChi Key: —
SMILES: c1cc[c-]c1.[C-]12[C-]3[C-]4[C-]5[C-]1[Mn]2345

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results