accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSOO(S)NH2OHO

CAS RN 7314-32-1

IUPAC Name: 2-amino-4-methanesulfonylbutanoic acid
Synonyms: methioninesulfone
Molecular Weight (g/mol): 181.21
Molecular Formula: C5H11NO4S
InChi Key: UCUNFLYVYCGDHP-UHFFFAOYNA-N
SMILES: CS(=O)(=O)CCC(N)C(O)=O
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)