accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7314-85-4

IUPAC Name: 1-bromoadamantane
Synonyms: 1-bromoadamantane
Molecular Weight (g/mol): 215.13
Molecular Formula: C10H15Br
InChi Key: VQHPRVYDKRESCL-UHFFFAOYSA-N
SMILES: BrC12CC3CC(CC(C3)C1)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results