accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 732-11-6

IUPAC Name: O,O-dimethyl {[(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)methyl]sulfanyl}phosphonothioate
Synonyms: percolate caswell no. 543 imidan 5 dust phosmet fosmet phtalofos stauffer R 1504 PMP safidon phosmet (substance)
Molecular Weight (g/mol): 317.31
Molecular Formula: C11H12NO4PS2
InChi Key: LMNZTLDVJIUSHT-UHFFFAOYSA-N
SMILES: COP(=S)(OC)SCN1C(=O)C2=CC=CC=C2C1=O
Molecular Weight (g/mol) 
  • (5)
  • (4)
Solvent or Matrix 
  • (3)
  • (2)
  • (1)
Concentration 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (5)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)