accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 73219-91-7

IUPAC Name: 3,5-dichloro-2,6-dimethoxybenzoate
Synonyms: 3,5-dichloro-2,6-dimethoxybenzoate
Molecular Weight (g/mol): 250.05
Molecular Formula: C9H7Cl2O4
InChi Key: JPIAALCEQSLBKF-UHFFFAOYSA-M
SMILES: COC1=C(C([O-])=O)C(OC)=C(Cl)C=C1Cl
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results