accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 73220-03-8

IUPAC Name: hydrogen 3-bromo-2,6-dimethoxy-N-[(1S)-1-(pyrrolidin-2-yl)propyl]benzamide chloride
Synonyms: hydrogen 3-bromo-2,6-dimethoxy-N-[(1S)-1-(pyrrolidin-2-yl)propyl]benzamide chloride
Molecular Weight (g/mol): 407.73
Molecular Formula: C16H24BrClN2O3
InChi Key: GYLIXNFLPDXSTM-YLIVSKOQSA-N
SMILES: [H+].[Cl-].CC[C@H](NC(=O)C1=C(OC)C=CC(Br)=C1OC)C1CCCN1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results