CAS RN 73220-39-0
IUPAC Name:
6-chloro-3-iodo-4H-chromen-4-one
Synonyms:
6-chloro-3-iodochromen-4-one
Molecular Weight (g/mol):
306.48
Molecular Formula:
C9H4ClIO2
InChi Key:
GMONZFZWCHBAKU-UHFFFAOYSA-N
SMILES:
ClC1=CC=C2OC=C(I)C(=O)C2=C1
1
–
1
of
1
results
1
6-Chloro-3-iodo-4H-chromen-4-one, 97%, Thermo Scientific™
CAS: 73220-39-0 Molecular Formula: C9H4ClIO2 Molecular Weight (g/mol): 306.483 MDL Number: MFCD09475515 InChI Key: GMONZFZWCHBAKU-UHFFFAOYSA-N Synonym: 6-chloro-3-iodochromone,6-chloro-3-iodo-4h-chromen-4-one,acmc-20alzd,6-chloro-3-iodo-chromone,6-chloro-3-iodo-4h-benzopyran-4-one,3-iodo-6-chloro-4h-1-benzopyran-4-one PubChem CID: 11666677 IUPAC Name: 6-chloro-3-iodochromen-4-one SMILES: C1=CC2=C(C=C1Cl)C(=O)C(=CO2)I