accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3HNH3COOCH3OCH3OOCH3

CAS RN 73257-47-3

IUPAC Name: 3,5-dimethyl 4-(4-methoxyphenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Synonyms: 3,5-dimethyl 4-(4-methoxyphenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Molecular Weight (g/mol): 331.37
Molecular Formula: C18H21NO5
InChi Key: IAXDEFZXLVTHLU-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)NC(C)=C(C1C1=CC=C(OC)C=C1)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results