CAS RN 7327-87-9
IUPAC Name:
1,4-dihydrazinylphthalazine; sulfuric acid
Synonyms:
dihydralazine sulfate; sulfuric acid
Molecular Weight (g/mol):
288.28
Molecular Formula:
C8H12N6O4S
InChi Key:
BWHAMWGGORIDBK-UHFFFAOYSA-N
SMILES:
OS(O)(=O)=O.NNC1=C2C=CC=CC2=C(NN)N=N1
1
–
2
of
2
results
1
Thermo Scientific Chemicals Dihydralazine sulfate
CAS: 7327-87-9 Molecular Formula: C8H10N6·H2O4S
2
Ophthazin Sulfate, TRC
CAS: 7327-87-9 Molecular Formula: C8 H10 N6 . H2 O4 S Molecular Weight (g/mol): 288.28 Synonym: Phthalazine, 1,4-dihydrazinyl-, sulfate (1:1),1,4-Phthalazinedione, 2,3-dihydro-, dihydrazone, sulfate (1:1) (9CI),Phthalazine, 1,4-dihydrazino-, sulfate (1:1) (8CI),1,4-Dihydrazinophthalazine sulfate,Depressan,Dihydralazine sulfate,Dihydrazinophthalazine sulfate,Dihyzin IUPAC Name: (Z)-[(4Z)-4-hydrazinylidene-2,3-dihydrophthalazin-1-ylidene]hydrazine;sulfuric acid SMILES: N\N=C\1/NN\C(=N/N)\c2ccccc12.OS(=O)(=O)O