accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH3CH3CH3CCH3SOOO

CAS RN 733-44-8

IUPAC Name: tetraethylazanium 4-methylbenzene-1-sulfonate
Synonyms: tetraethylammonium tosylate
Molecular Weight (g/mol): 301.45
Molecular Formula: C15H27NO3S
InChi Key: QKFFSWPNFCXGIQ-UHFFFAOYSA-M
SMILES: CC[N+](CC)(CC)CC.CC1=CC=C(C=C1)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)