accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OONO

CAS RN 73365-02-3

IUPAC Name: tert-butyl 2-formylpyrrolidine-1-carboxylate
Synonyms: tert-butyl 2-formylpyrrolidine-1-carboxylate
Molecular Weight (g/mol): 199.25
Molecular Formula: C10H17NO3
InChi Key: YDBPZCVWPFMBDH-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCCC1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results