CAS RN 73401-74-8
IUPAC Name:
2-[4-(hydroxymethyl)phenyl]acetic acid
Synonyms:
[4-(hydroxymethyl)phenyl]acetic acid
Molecular Weight (g/mol):
166.18
Molecular Formula:
C9H10O3
InChi Key:
FWZBPBKAANKOJQ-UHFFFAOYSA-N
SMILES:
OCC1=CC=C(CC(O)=O)C=C1
1
–
1
of
1
results
1
4-(Hydroxymethyl)phenylacetic acid, 97%
CAS: 73401-74-8 Molecular Formula: C9H10O3 Molecular Weight (g/mol): 166.176 MDL Number: MFCD00065692 InChI Key: FWZBPBKAANKOJQ-UHFFFAOYSA-N Synonym: 4-hydroxymethyl phenylacetic acid,2-4-hydroxymethyl phenyl acetic acid,4-hydroxymethyl phenyl acetic acid,4-hydroxymethylphenylacetic acid,benzeneacetic acid,4-hydroxymethyl,benzeneacetic acid, 4-hydroxymethyl,ambotzrl-1037,pubchem19630,acmc-1bl5i PubChem CID: 3283498 IUPAC Name: 2-[4-(hydroxymethyl)phenyl]acetic acid SMILES: C1=CC(=CC=C1CC(=O)O)CO