accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HISnHH3CH3CH3C

CAS RN 7342-47-4

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 418.98
Molecular Formula: C12H29ISn
InChi Key: PGQGYULKMYCHJV-UHFFFAOYSA-N
SMILES: I.CCCC[SnH](CCCC)CCCC
Boiling Point 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results