accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OH2OOClOOOOClOOOHg

CAS RN 73491-34-6

IUPAC Name: mercury(2+) trihydrate diperchlorate
Synonyms: mercury(2+) trihydrate diperchlorate
Molecular Weight (g/mol): 453.53
Molecular Formula: Cl2H6HgO11
InChi Key: DFJWPGPUWHLJIJ-UHFFFAOYSA-L
SMILES: O.O.O.[Hg++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results